Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acid Reagent Bottle Holder
Strong and stable.
Supplier: Lab Workx LLC 9087
This item is not returnable.
View return policy
Description
- Wooden tray with space for five 1.01 oz. (30mL) reagent bottles
- Heavy enough to maintain stability whether bottles are filled or empty
- Sufficient clearance to provide paper lining which may be replaced from time to time
Order Info
Bottles not included; Dimensions: 8 x 2.8 x 2 in. (20 x 7 x 5cm)
Specifications
Acid Reagent Bottle Holder | |
Wood | |
8 x 2.8 x 2 in. (20 x 7 x 5cm) | |
8 in. | |
2.8 in. | |
2 in. |
Not Autoclavable | |
5 | |
20 cm | |
7 cm | |
5 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction