Learn More
Newcomer Supply Acid Fast Bacteria (AFB), Animal Control Slides, 98/set
Supplier: Newcomer Supply 4011B
Newcomer Supply Acid Fast Bacteria (AFB), Animal Control Slides are for the positive histochemical staining of acid-fast mycobacteria in tissue sections. Acid Fast Bacteria (AFB), Animal Control Slides contain sections of positive staining animal organ and negative staining human lung.