Learn More
Fisher Science Education™ Upright Compound Microscope Achromat Objective
Achromat Upright Microscope Objective with correction for color and spherical aberration.
$17.50 - $40.75
Specifications
Type | Achromat Objective |
---|---|
Autoclavable | Not Autoclavable |
Length (Metric) | 160 mm |
Sterility | Non-sterile |
Voltage | 0/1 V |
Description
- RMS
- DIN standard
- Available in 4X or 40X
- Achromat Microscope Objective
- Correction for color and spherical aberration
Specifications
Achromat Objective | |
160 mm | |
0/1 V |
Not Autoclavable | |
Non-sterile | |
5 years optical and mechanical, 1 year electrical from date of purchase |