Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone, ULTRA RESI-ANALYZED, J.T. Baker™
Click to view available options
Quantity:
4 L
Specifications
Melting Point | -94.7°C |
Color | Colorless |
Quantity | 4 L |
Flash Point | −20°C |
Physical Form | Liquid |
Titratable Base | ≤ 0.6(μeg/g) |
UN Number | UN 1090 |
Water | ≤0.5% |
Formula Weight | 58.08 |
Percent Purity | ≥99.4% |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | Hazard Statement: Highly flammable liquid and vapor. Causes serious eye irritation. May cause drowsiness or dizziness. May be harmful if swallowed and enters airways. |
Precautionary Statement | Precautionary Statement In case of fire: Use water spray, foam, dry powder or carbon dioxide for extinction. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove person to fresh air and keep comfortable for breathing. Call a POISON CENTERPrevention: Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking. Keep container tightly closed. Use explosion-proof electrical/ventilating/lighting equipment. Ground and bond container and receiving equipment. Use only non-sparking tools. Take precautionary measures against static discharge. Wash thoroughly after handling. Avoid breathing dust/mist/vapors/spray. Use only outdoors or in a well-ventilated room. |
Recommended Storage | Store in a well-ventilated place. Keep cool. Store locked up. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction