Learn More
Ace Glass, Inc. Tube, pressure, 9mL, 150psi, 19mm OD, 10.2cm long, complete with#7 front-seal plug
SDP
Supplier:  Ace Glass, Inc. 864817
Encompass_Preferred
9mL glass pressure tube with #7 Ace-Thred, rated at 150psig at as much as 120C. Includes #7 front-seal plug. Dimensions: 19mm O.D. x 10.2cm long.
Catalog No. NC9198633