Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Ace Large Chromatography Column, 600 mm

Suitable for ion-exchange or activated-carbon work. Order column adapters separately.

Supplier:  MilliporeSigma™ Supelco™ Z1801061EA

 View more versions of this product

Catalog No. 11-100-7153


Add to Cart

Description

Description

Rugged borosilicate glass column with No. 50 internal Ace-Threds on ends to which a wide variety of upper and lower adapters can be fitted.
Specifications

Specifications

75 mm
Ace
Chromatography
600 mm
2.65 L
Borosilicate Glass
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit