Learn More
Fisherbrand™ accumet™ Portable AP110 pH Meter
For accurate measurements anytime, anywhere.
Includes: Meter, 9V battery, and manual.
Description
- Large LCD with backlighting and plain-language text
- Rugged waterproof IP67 housing
- 200 data point internal memory
pH Mode
- Range: -2.00 to +20.00
- Resolution: 0.1/0.01
- Accuracy: ±0.01
- Calibration: Up to five or six points from threesets: (USA: 2.00, 4.00, 7.00, 10.00 12.00; NIST: 1.68, 4.01, 6.86, 9.18, 12.45; EURO: 1.00, 3.00, 6.00, 8.00, 10.00, 13.00)
Warranty and Services
Three year meter warranty, one year electrode warranty
Specifications
Meter, 9V battery, and manual | |
pH reading | |
Rugged waterproof IP67 housing | |
AP110 pH Meter |