Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Germaine™ Laboratories AccuHome™ Pregnancy Test
Germaine™ Laboratories AccuHome™ Pregnancy Test is designed for self testing.
$7.75 - $11.25
Specifications
Content And Storage | 15°C to 30°C |
---|---|
Clia Complexity | Waived |
Detectable Analytes | hCG |
Sample Type | Urine |
Sensitivity | 25 mIU/mL |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-111-349
|
Germaine™ Laboratories
89102 |
2 Tests |
Pack of 2 for $11.25
|
|
|||||
23-111-350
|
Germaine™ Laboratories
89101 |
1 Test |
Each for $7.75
|
|
|||||
Description
- CLIA Complexity: Waived
- Test results within three minutes
- Combination of monoclonal and polyclonal antibody reagent selectively detects elevated levels of human chorionic gonadotropin (hCG)
- Positive specimens react with the specific antibody-hCG colored conjugate and form a colored line at the T (test) area
- Absence of this colored line suggests a negative result
- A colored line at the C (control) area will always appear regardless of the presence or absence of hCG
Specifications
15°C to 30°C | |
hCG | |
25 mIU/mL | |
3 min. |
Waived | |
Urine | |
Yes |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title