Learn More
Hemosure™ Accu-Reader™ A100 Analyzer
The Hemosure™ Accu-Reader™ A100 is a fully automated FIT system intended for the in vitro qualitative detection of fecal occult blood in feces.
Supplier: Hemosure™ AR01M
Description
- Automated Fecal Immunochemical Test (FIT) Analyzer
- Accurate detection of fecal occult blood
- Easy-to-use touchscreen
Specifications
Touchscreen |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'