Learn More
IMEB, Inc Sakura Accu-Edge Low Profile Microtome Blades, Disposable, 50 per Pack
Supplier: IMEB, Inc 4689
The Accu-Edge disposable low-profile blades by Sakura Finetek are compatible with all Accu-Edge blade holders.This product is ideal for microtomy, cryotomy, and sectioning specimens without striations, distortion, or chattering. It is sold in dispensers that can store 50 blades and distribute them in a few seconds. The previously used blade is then automatically ejected. 50 per pack.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.