Learn More
Medex Supply Sakura Accu-Edge 4689 Microtome Blade, Low Profile, Disposable, 50/Pk
Supplier: Medex Supply SAKM732141
Accu-Edge disposable microtome blades deliver excellent results in the world of microtomy. The Accu-Edge disposable microtome blades can section down to one micron without striation, compression, distortion, or chattering.
This item is not returnable.
View return policy