Learn More
Scott Safety™ Accessory For SEMS II Personnel Accountability System: RFID Programmer
For use with the SEMS II system
$178.15 - $937.84
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-823-812
|
Scott Safety™
7012471568 |
RFID Card |
Pack of 10 for $178.15
|
|
|||||
19-823-811
|
Scott Safety™
7012471567 |
RFID Card Programmer with Software |
Each for $937.84
|
|
|||||
19-823-813
|
Scott Safety™
7012471994 |
Sems II Repeater | N/A | N/A | |||||
Please contact your local Fisher Scientific sales representative for pricing | |||||||||
Description
Scott's SEMS II is a remarkable accountability system that puts the incident commander at every firefighter's side, displaying information vital to personal safety in volatile and dangerous situations, while providing a continuous link of communications to the command station. And with the integrated personnel locator, it provides the peace of mind that a downed or trapped firefighter can be quickly found.
RFID / Personalized Tagging. The system can be personally customized and programmable using RFID identification cards to link a firefighter to a specific Air-Pak SCBA. To enter their personal information, each firefighter simply swipes their pre-programmed ID card on the back of the SEMS II console. This information is transmitted to the base command computer while SEMS II is activated. Each card has three programmable fields for firefighter identification that are programmed via an RFID tag writer and proprietary software.
RFID Cards are used to link a firefighter to specific Air-Pak SCBA.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.