Learn More
Conair™ Accessories and Replacements for Waring™ Blenders::Switch for Hazardous Location Blender Motor
Supplier: Conair™ BHLSW
Description
Use with 14-509-31 Blender Motor. For 115VAC, 8A. UL listed for Class 1, Group D hazardous locations.
Specifications
Foot Switch | |
Waring™ blender motor |