Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces Access Cubby with White Metal Shelves
Cubby units available in both oak and maple veneers, and in sections from one to four. The sections are divided with fixed or adjustable 16-gauge metal shelvingn. The shelves are powder coated with a matte finish for a modern look. The back is designed to be anchored to a wall for stability.
$1721.00 - $7765.00
Specifications
For Use With (Application) | Storage |
---|---|
Includes | Metal Shelves |
No. of Drawers | None |
Color | White |
Catalog Number | Mfr. No. | Material | No. of Shelves | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | No. of Shelves | Width (Metric) | Price | Quantity & Availability | |||
S14398
|
Diversified Spaces
CC541215WM |
Maple | 3 | 30.48 cm |
Each for $1,721.00
|
|
|||
S14390
|
Diversified Spaces
CC781215WM |
Maple | 5 | 30.48 cm |
Each for $2,503.00
|
|
|||
S14396
|
Diversified Spaces
CC542415WM |
Maple | 6 | 60.96 cm |
Each for $2,617.00
|
|
|||
S14489
|
Diversified Spaces
CC543615WM |
Maple | 9 | 91.44 cm |
Each for $3,690.00
|
|
|||
S14378
|
Diversified Spaces
CC782415WM |
Maple | 10 | 60.96 cm |
Each for $3,945.00
|
|
|||
S14434
|
Diversified Spaces
CC544815WM |
Maple | 12 | 121.92 cm |
Each for $4,759.00
|
|
|||
S14418
|
Diversified Spaces
CC783615WM |
Maple | 15 | 91.44 cm |
Each for $5,636.00
|
|
|||
S14399
|
Diversified Spaces
CC784815WM |
Maple | 20 | 121.92 cm |
Each for $7,324.00
|
|
|||
S14438
|
Diversified Spaces
CC541215WK |
Oak | 3 | 30.48 cm |
Each for $1,857.00
|
|
|||
S14440
|
Diversified Spaces
CC542415WK |
Oak | 6 | 60.96 cm |
Each for $2,793.00
|
|
|||
S14495
|
Diversified Spaces
CC782415WK |
Oak | 10 | 60.96 cm |
Each for $4,191.00
|
|
|||
S14479
|
Diversified Spaces
CC544815WK |
Oak | 12 | 121.92 cm |
Each for $5,074.00
|
|
|||
S14496
|
Diversified Spaces
CC783615WK |
Oak | 15 | 91.44 cm |
Each for $5,977.00
|
|
|||
S14484
|
Diversified Spaces
CC784815WK |
Oak | 20 | 121.92 cm |
Each for $7,765.00
|
|
|||
Description
- Choose from oak or maple hardwood and veneers with an earth-friendly UV finish
- Cubbies have an adjustable height
- Finished with rubber base molding, metal corners and durable chemical-resistant UV finish
- Made in the U.S.A.
- Metal shelving powder coated
- Sections are divided with fixed or adjustable 16-gauge metal shelving, featuring a diamond pattern
- The back is designed to be anchored to a wall for stability
Specifications
Storage | |
Metal Shelves | |
None | |
White |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title