Learn More
Biotium Ac-IETD-R110
Supplier: Biotium 10207
Z-IETD-R110 is a substrate for caspase-8, which is involved at the early stage of apoptosis, acting as an initiator of the caspase activation cascade.Properties: Full name: Rhodamine 110, bis-(N-acetyl-L-isoleucyl-L-glutamyl-L-threonyl-L-aspartic acid amide; Ex/Em of end product (R110) = 496/520 nm; Off-white to pink solid soluble in DMSO; Store at -20°C and protect from light; C68H82N10O24; MW: 1422
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.