Learn More
MilliporeSigma ABSORPTION CUVETTES STND CELLS
Supplier:  MilliporeSigma Z8032271EA
Encompass
High Performance Quartz Glass, spectral range 200-2500 nm, pathlength 2 mm, chamber volume 700 μL
Catalog No. NC2389340