Learn More
New Pig Absorb-and-Lock™ Water Absorbent Strip
Deploy these super absorbent strips to absorb high volume leaks and drips in problem areas.
Supplier: New Pig WTR001
This item is not returnable.
View return policy
Description
- Super Absorbent Polymer (SAP) filler swells on contact with water and expands to pull in large volumes of liquid quickly
- Absorbed water is locked inside; won't leach or run out when you pick up or dispose, even when fully saturated
- Water locking gel mat strips are ideal for sewage and restroom backups and overflows and at the base of equipment, appliances and machinery
- Expanded strips create a barrier to help prevent further leaks and drips from escaping and spreading into walkways; also good to help stop water from seeping under doors
- Strips (before water contact) are low-profile to fit tight spaces; unobtrusive design won't create trip hazards
- Leave strip down until fully saturated, then dispose
Specifications
Polypropylene |