Learn More
Thermo Scientific™ ABsolute QPCR Mix, SYBR Green, fluorescein
Optimized for SYBR Green chemistry and contains all the components necessary to perform quantitative PCR, with the exception of template and primers.
Supplier: Thermo Scientific™ AB1220A
Description
The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets.
ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, Fluorescein or no reference dye. A master mix optimized for capillary instruments is also available.
- Thermo Scientific ABsolute qPCR SYBR Green Mixes are optimized for SYBR Green chemistry and contain all the components necessary to perform quantitative PCR, with the exception of template and primers
- The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets
- ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, low ROX concentration, fluorescein or no reference dye
- A master mix optimized for capillary instruments is also available
Highlights
- High Sensitivity & Specificity – The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
- Optimised buffer containing SYBR Green I
- Reproducibility – All master mixes undergo extensive QC to ensure batch to batch consistency
- Store at -20°C
Specifications
Contains: • 1 x 5 mL ABsolute QPCR SYBR Green Fluorescein Mix (2X) •100 μL MgCl2 (1M) Sufficient for 400 x 25 μL reactions. Store at -20°C |
|
High | |
Taq DNA Polymerase | |
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5 | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
2X | |
DNA, dsDNA |
SYBR | |
qPCR | |
Standard | |
ABsolute | |
400 Reactions of 25 μL Each | |
qPCR Cyclers | |
400 Reactions | |
5 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.