Learn More
Andwin Scientific ABS PARAFILM DISPENSER BL EA
SDP
Supplier:  Andwin Scientific 7099301
Encompass_Preferred
The ABS Parafilm Dispenser features a unique design that provides maximum user safety. This product can store, dispense, and cut Parafilm, tape, labels, and Tough-Tags. The dispenser has a safety razor that slides and cuts with no danger to your fingers. It is able to store one or two 2-inch rolls, or one 4-inch roll.
Catalog No. 50-727-499