Learn More
Novus Biologicals™ Abpure™ TCS Antibody Purification Kit
Novus Biologicals™ Abpure TCS Antibody Purification Kit can be used in its rapid version by spin-purification in centrifuge, or by gravity-flow purification.
$542.50 - $1183.00
Description
- Fully compatible with InnovaCoat GOLD and LATEX conjugation kits which allow purified antibody to be conjugated rapidly
- Antibody to be purified should be in 10mL to 50mL of tissue culture supernatant (TCS)
- Up to 5mg of antibody can be purified in each run
Specifications
4°C | |
Antibody Purification Kit |
For research use only.