Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ CytoQuest™ Epithelial Cell Kit
Contains antibodies for immobilization and immunostaining of circulating epithelial cells
Supplier: Abnova™ KA4439
Includes:
- Anti-EpCAM capturing antibody (Biotin): Biotin conjugated Anti-EpCAM antibody for CTC capturing.
- Anti-PanCK detecting antibody (FITC): FITC conjugated Anti-PanCK antibody for CTC detection.
- Anti-CD45 detecting antibody (PE): PE conjugated Anti-CD45 antibody for CTC detection.
- 50X Antibody Dilution Buffer (50X ADB)
Description
- For use with CytoQuest CR
- Includes reagents that are sufficient for 20 assays using recommended protocol
Immunofluorescence (Circulating Tumor Cell)
Recommended Storage
Store Anti-EpCAM capturing antibody (Biotin), Anti-PanCK detecting antibody (FITC) and Anti-CD45 detecting antibody (PE) at 4°C. Store 50X Antibody Dilution Buffer (50X ADB) at -20°C. Aliquot to avoid repeated freezing and thawing.
Specifications
CytoQuest CR | |
|
|
1 Kit | |
Store Anti-EpCAM capturing antibody (Biotin), Anti-PanCK detecting antibody (FITC) and Anti-CD45 detecting antibody (PE) at 4°C. Store 50X Antibody Dilution Buffer (50X ADB) at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only.
Spot an opportunity for improvement?
Provide Content Correction