Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ CytoQuest™ CR: SCx™ Spiral Chamber
Single use, replaceable sample chamber to avoid cross contamination on CytoQuest CR
Supplier: Abnova™ U0314
Includes: Set contains 20 pieces each of: Coil holder, connecting base
Description
Applications:Immunofluorescence
Recommended Storage
Store at room temperature.
Specifications
CytoQuest CR | |
Store at room temperature. | |
Set contains 20 pieces each of: Coil holder, connecting base | |
1 Set |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only (RUO).