Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ CytoQuest™ CR: CytoChipNano
CytoChipNano is a single use, replaceable chip for capture, enumeration, and isolation of CRCs on CytoQuest CR
Supplier: Abnova™ U0095
Includes: Each set contains 20 CytoChipNano.
Description
Applications:Immunofluorescence
Recommended Storage
Store at 4°C. Avoid repeated freezing and thawing.
Specifications
CytoQuest CR | |
Store at 4°C. Avoid repeated freezing and thawing. | |
Each set contains 20 CytoChipNano. | |
1 Set |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For research use only (RUO).
Spot an opportunity for improvement?
Provide Content Correction