Learn More
Radiometer America Radiometer Inlet Gasket with Holder for ABL90 FLEX PLUS
Supplier: Radiometer America 903585
ABL90 Inlet Gasket with Holder (For ABL90 FLEX PLUS)
Inlet Gasket Holder for the ABL90 FLEX PLUS blood gas analyzer.
ABL90 FLEX PLUS Blood Gas Analyzer
The ABL90 FLEX PLUS analyzer gives you reliable results. With the ABL90 FLEX PLUS analyzer you get lab quality test results. In just 35 seconds, our cassette-based ABL90 FLEX analyzer can deliver 17 parameters from a sample as small as 65 μL. ABL90 measures blood gas, electrolytes, metabolites and co-oximetry. The ABL90 analyzer can measure a sample from a syringe, capillary tube or test tube.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.