Learn More
Anton Paar Digital Compact Refractometer – Abbemat 3200
Supplier:  Anton Paar 173453
Encompass_Preferred
The Abbemat 3200 refractometers embody reliability, precision, and convenience. These benchtop refractometers offer you an exceptional combination of technical expertise and user-friendly operation. Use the in-built application knowhow to determine the concentration of binary solutions. With over 200 available methods – from acetic acid over to soft-drinks and up to Zeiss – Abbemat refractometers provide the quality control that you require.- For all concentration measurements e.g. acc. to AOAC and for iodine number determination- Automated data transfer via LIMS for maximum data storage- Measure all liquid samples with the largest refractive index rangeMeasuring range Refractive index scale (RI)- Range nD 1.30 to 1.72 - Resolution nD ± 0.0001- Accuracy nD ± 0.0001Measuring range Brix scale- Range 0 to 100 %- Resolution 0.01 %- Accuracy 0.05 %Temperature- Range 15 to 60 °C - Accuracy ± 0.05 °C
Catalog No. NC1818327