Learn More
Amsbio LLC AAV Testing Kit. Contains CMV-GFP in AAV serotypes 1, 2, 5, 6, 8, 9, and DJ. Titer 1x10^13 GC/mL. 50 ul each.
Supplier: Amsbio LLC CT0002
AAV Testing Kit. Contains CMV-GFP in AAV serotypes 1, 2, 5, 6, 8, 9, and DJ. Titer 1x10^13 GC/mL. 50 ul each.