Learn More
Perkin Elmer US LLC Single Slot Burner Head for PerkinElmer AAnalyst Series Spectrometers, Slot Length: 5cm, Flame Type: Nitrous Oxide-Acetylene
Supplier: Perkin Elmer US LLC N0400100
The 5 cm nitrous oxide burner head is required for nitrous oxide-acetylene operation. This burner head can also be used with air-acetylene or rotated to provide reduced sensitivity.