Learn More
Perkin Elmer US LLC AA Ultra Quiet Oil-Less Air Compressor, 220V/50-60Hz
Supplier: Perkin Elmer US LLC N9306346
Features and Benefits: Completely quiet oil-less operation Delivers clean and dry instrument grade air Water separator with 5 micron particle filter assures no liquid transfer Specialty coated cylinders and high temperature piston rings for long service life Durable air tank with automatic condensate drain, internal coating and 5 year warranty Low annual operating costs 2-year system warranty A complete compressed air station incorporating an outlet pressure regulator, tank and outlet pressure gauge, shut off ball valve, ASME safety valve, and pressure switch allowing for complete automatic operation Specifications: Output CFM: 4.2 CFM Output L: 120 L/min Max Pressure: 120 PSI Tank Size: 24 L Dimensions: 16 in x 16 in x 21 in
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.