Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGuard™ A31A1 Series Powder-Free Latex 5.5 mil Exam Gloves, Cream
SDP

Catalog Number p-7239906
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

A31 Cream Latex Exam Gloves provide all purpose protection for medical use. Latex material fits around hand for superior comfort and dexterity.

  • Latex
  • Powder-free
  • Non-Sterile
  • Ambidextrous
  • Exam Grade
  • Color: Cream
  • Texture: Finger and Palm
  • Palm Thickness: 4.5 mil
  • Fingertip Thickness: 5.5 mil
  • Length: Standard (23 cm)
  • Cuff: Beaded
  • 100 Gloves/Box
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Cream
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 5.5 mil
Latex Free No
Material Latex
Palm Thickness (English) 4.5 mil
Powdered Powder Free
Size Small
Sterility Non-sterile
Thickness (English) 5.5 mil
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model A31A11
Series Vanguard Safety
Length (Metric) 23 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit