Learn More
SANITARY FITTINGS A 2" SANITARY SIGHT GLASS, 5.8
SDP
Supplier:  SANITARY FITTINGS 54BMP2316
Encompass_Preferred
A 2" sanitary sight glass, 5.875" long, made from borosilicate glass and 316 stainless steel
Catalog No. NC2144666