Learn More
Thermo Scientific™ Oxoid™ IMAGEN™ Respiratory Syncytial Virus (RSV) Positive Control Slide
Oxoid IMAGEN Respiratory Syncytial Virus Positive Control is a slide with acetone-fixed human epithelial cells infected with RSV.
Supplier: Thermo Scientific™ S6108302
Description
The IMAGEN™ Control Slides are for use as a control to the following tests: IMAGEN Adenovirus (K610011-2) IMAGEN Respiratory Syncyti al Virus (RSV) (K610211-2) IMAGEN Parainfluenza Group (K610311-2) IMAGEN Parainfluenza Typing 1, 2 and 3 (K610411-2) IMAGEN Influenza A and B Virus (K610511-2) IMAGEN Respiratory Screen (K612011-2) IMAGEN hMPV (K612511-2) IMAGEN Herpes Simplex Virus (HSV) (K610611-2) IMAGEN Chlamydia kit (K610111-2)
Sealed in foil pouch filled with nitrogen.Specifications
IMAGEN™ Respiratory Syncytial Virus (RSV) Positive Control Slide | |
Control | |
1 Slide/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.