Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD13 Rat anti-Mouse, RY586, Clone: R3-242, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 753222
This item is not returnable.
View return policy
Description
CD13 Monoclonal antibody specifically detects CD13 in Mouse samples. It is validated for Flow CytometrySpecifications
CD13 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 |
Flow Cytometry | |
R3-242 | |
RY586 | |
Anpep; Alanyl aminopeptidase; Aminopeptidase N; AP-N; mAPN; Lap-1 | |
Crude Aminopeptidase N Preparation from BALB/c Mouse Intestinal Epithelium | |
RUO | |
Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction