Learn More
Thermo Scientific™ 9mm Certified Clear Wide Opening Screw Thread Kits
Use certified clean, low particle, low background vials for your high sensitivity LC/MS and GC/MS applications. They are fully lot-tested for 15 critical parameters.
$84.00 - $403.50
Specifications
Color Clear
No. per Pack 100
View More Specs
Products 25
Catalog Number Mfr. No. Description Closure Color Septum Marking Spot Price Quantity & Availability  
Catalog Number Mfr. No. Description Closure Color Septum Marking Spot Price Quantity & Availability  
03-060-042
View Documents
Thermo Scientific™
CERT500071LV
Convenience Kit, Certified 9 mm Screw Vial, 350uL Fused Insert, with bonded PTFE/Silicone Slit Cap Blue Bonded Red/PTFE White Silicone Pre-slit Yes
Pack of 100 for $240.50
Add to cart
03-060-034
View Documents
Thermo Scientific™
CERT500078
Convenience Kit, Certified 9 mm Screw Clear Vial 2mL with bonded PTFE/Silicone Cap Blue Bonded Red/PTFE White Silicone No
Pack of 100 for $100.80
Add to cart
03-151-692
View Documents
Thermo Scientific™
CERT5000580W
Convenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL Blue Ivory PTFE/Red Rubber Yes
Pack of 100 for $84.00
Edge
Add to cart
03-377-295
View Documents
Thermo Scientific™
CERT5000578W
Convenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL Blue Red PTFE/White Silicone Yes
Pack of 100 for $100.80
Add to cart
03-060-045
View Documents
Thermo Scientific™
CERT500093
Convenience Kit, Certified 9 mm Screw Clear Vial 2mL with bonded PTFE/Silicone Cap Pre-slit Blue Bonded Red/PTFE White Silicone No
Pack of 100 for $112.40
Add to cart
03-052-450
View Documents
Thermo Scientific™
CERT5000593W
Convenience Kit, Certified 9mm Wide Opening SureStop Vial, 2mL Blue Blue PTFE/White Silicone Yes
Pack of 100 for $146.00
Edge
Add to cart
03-060-048
View Documents
Thermo Scientific™
MSCERT5000134W
Assembled Convenience Kit, Certified 9 mm Screw Clear I-D Vial 2 mL with bonded PTFE/Silicone Cap Blue Bonded White Silicone/Clear PTFE MS Certified Yes
Pack of 100 for $185.90
Add to cart
03-050-593
View Documents
Thermo Scientific™
CERT500080W
Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL Blue Ivory PTFE/Red Rubber Yes
Pack of 100 for $91.40
Add to cart
03-050-596
View Documents
Thermo Scientific™
CERT500092W
Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL Blue Red PTFE/White Silicone Yes
Pack of 100 for $96.60
Edge
Add to cart
03-060-053
View Documents
Thermo Scientific™
MSCERT500033TR
Convenience Kit, 9mm Wide Opening 1.5mLTotal Recovery Screw Vial with 10μL Reservoir Blue Bonded Clear PTFE/Clear Silicone No
Pack of 100 for $356.00
Add to cart
03-060-054
View Documents
Thermo Scientific™
MSCERT5000341W
Convenience Kit, 9mm Wide OpeningScrew Vial, 2mL Blue Bonded Clear PTFE/Clear Silicone Yes
Pack of 100 for $116.60
Edge
Add to cart
03-060-060
View Documents
Thermo Scientific™
MSCERT500038
Convenience Kit, 9mm Wide Opening 1.7mLHigh Recovery Screw Vial with 30μL Reservoir Gray Bonded Clear PTFE/Clear Silicone No
Pack of 100 for $266.00
Add to cart
03-060-062
View Documents
Thermo Scientific™
MSCERT5000401W
Convenience Kit, 9mm Wide OpeningScrew Vial, 2mL Gray Bonded Clear PTFE/Clear Silicone Yes
Pack of 100 for $130.20
Add to cart
03-060-056
View Documents
Thermo Scientific™
MSCERT5000S34W
Convenience Kit, 9mm Wide OpeningScrew Vial, 2mL, Silanized Blue Bonded Clear PTFE/Clear Silicone Yes
Pack of 100 for $125.00
Add to cart
03-060-064
View Documents
Thermo Scientific™
MSCERT5000S40W
Convenience Kit, 9mm Wide OpeningScrew Vial, 2mL, Silanized Gray Bonded Clear PTFE/Clear Silicone Yes
Pack of 100 for $129.20
Add to cart
03-060-039
View Documents
Thermo Scientific™
CERT5000193W
Assembled Kit, Certified 9mm Wide Opening Screw Vial, 2mL Gray Bonded Red PTFE/White Silicone Yes
Pack of 100 for $130.20
Add to cart
03-060-027
View Documents
Thermo Scientific™
CERT500078W
Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL Blue Bonded Red PTFE/White Silicone Yes
Pack of 100 for $142.80
Edge
Add to cart
03-060-044
View Documents
Thermo Scientific™
CERT500079
Convenience Kit, Certified 9mm Wide Opening High Recovery Screw Vial, 1.7mL Gray Bonded Clear PTFE/Clear Silicone No
Pack of 100 for $343.50
Add to cart
03-060-029
View Documents
Thermo Scientific™
CERT500093W
Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL Gray Bonded Clear PTFE/Clear Silicone Yes
Pack of 100 for $114.50
Edge
Add to cart
03-060-036
View Documents
Thermo Scientific™
CERT5000992
Convenience Kit, Certified 9mm Wide Opening High Recovery Screw Vial, 1.7mL Black Bonded Red PTFE/White Silicone No
Pack of 100 for $328.00
Add to cart
03-060-046
View Documents
Thermo Scientific™
CERT5000993
Convenience Kit, Certified 9mm Wide Opening Total Recovery Screw Vial, w/ 10μL Reservoir Gray Bonded Clear PTFE/Clear Silicone No
Pack of 100 for $403.50
Add to cart
03-060-043
View Documents
Thermo Scientific™
CERT500073LV
Convenience Kit, Certified 9 mm Screw Vial, 350uL Fused Insert, with bonded PTFE/Silicone Slit Cap Blue Bonded Red/PTFE White Silicone Pre-slit No This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
03-050-592
View Documents
Thermo Scientific™
CERT500080
Convenience Kit, Certified 9mm Wide Opening Screw Vial, 2mL Blue Ivory PTFE/Red Rubber No This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
03-060-052
View Documents
Thermo Scientific™
MSCERT500032
Convenience Kit, 9mm Wide Opening 1.7mLHigh Recovery Screw Vial with 30μL Reservoir Blue Bonded Clear PTFE/Clear Silicone No This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
03-060-061
View Documents
Thermo Scientific™
MSCERT500039TR
Convenience Kit, 9mm Wide Opening 1.5mLTotal Recovery Screw Vial with 10μL Reservoir Gray Bonded Clear PTFE/Clear Silicone No This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Use certified clean, low particle, low background vials for your LC/MS, GC/MS and chromatography high sensitivity applications. Thermo Scientific™ 9mm Wide Opening Screw Thread Certified Vial Kits are fully lot-tested by multiple methods for 15 critical parameters. Every pack comes with a Certificate of Conformance. Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, vials are available in assembled and unassembled kits. An assortment of wide opening 9mm vials and a selection of closures are available to suit your application.
Unassembled kits
  • 100 vials and 100 caps with pre-assembled septa
  • Reusable two-compartment trays protect vials and closures while keeping matching supplies together
  • Clear trays make it easy to keep track of available supplies without opening containers
Assembled kits
  • 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers
Specifications
Specifications
Clear
100
SDS
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.