Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotix™ 96-Well Format Assay Plates

Large well diameter with raised “Chimney”-style wells help reduce cross-contamination.

Supplier:  Biotix™ AP03509CU

 View more versions of this product

Catalog No. 12-111-072



Edge
Add to cart

Description

Description

  • Made from medical grade virgin polypropylene
  • DNase, RNase and Endotoxin (pyrogen) free
  • Alphanumeric grid to help with sample identification and organization
  • Notched bottom corner to be used as a reference for plate orientation
  • Available in V-shaped and U-shaped wells
Specifications

Specifications

No
Standard
8 x 12
Clear
Polypropylene
96
Non-treated
10
14.6 mm
U
Without Lid
Non Binding
Assay Plate
330 μL
Non-sterile
350 μL
85.48 mm
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit