Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab 95% Alcohol CytoVial Prefills
95% reagent alcohol prefills.
Supplier: StatLab CVP9500/100
Alert:
Harmful if swallowed, in contact with skin, or if inhaled
Description
- CytoVial Prefill 95% Alcohol 50 mL
- 100 per case
- Vials are pre-filled with 95% reagent alcohol and hold 4 slides
- Screw-top caps assure tight fit and leak resistance during transit
Specifications
Pre-filled Pap Jar | |
Plastic | |
Non-sterile | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Pre-filled container
Spot an opportunity for improvement?
Provide Content Correction