Learn More
WELCH MATERIALS INC DoprahTM 9-425 Blue Open Top Ribbed Screw Cap with 9 mm White PTFE/Red Silicone Septa 1.0mm Thick. 100pcs/pk, 10pk/cs
Supplier: WELCH MATERIALS INC DC11461
Cap&Septa: 9-425 Blue Open Top Ribbed Screw Cap with 9 mm White PTFE/Red Silicone Septa 1.0mm Thick. 100pcs/pk, 10pk/cs