Learn More
Ace Glass, Inc. 915mm x 14mm OD porosity B straight sparger tube, 1/2" connection top
Supplier: Ace Glass, Inc. 645345
915mm X 14mm O.D. straight sparger tube with a 70-100 micron fritted disc at the bottom, and a 1/2 I.D. hose connection tubing at the top. Will fit inside #15 Ace-Threds.