Learn More
Perkin Elmer US LLC 9 mm Pre-Assembled Blue Polypropylene Screw Cap with PTFE/Silicone Septa, 100/pk
Supplier: Perkin Elmer US LLC N9306360
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.
missing translation for 'provideContentCorrection'
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'