Learn More
Perkin Elmer US LLC 9 mm Pre-Assembled Blue Polypropylene Screw Cap with PTFE/Silicone Ultra-Low Bleed Septa, 100/pk
Supplier: Perkin Elmer US LLC N9306362
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.