Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TechNiGlove International Nitrile 9.5 in. XGloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
Specifications
Certifications/Compliance | FDA 29 CFR 1910, NFPA 1999, FDA 21 CFR 170-19, ISO 9001:2000, ISO 14001:2004 |
Applications | Laboratory, Industrial |
AQL | 1.5 |
Color | Blue |
Disposable | Yes |
Exterior Finish | Fully Textured |
Laboratory/Cleanroom Use | No |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Show More |