Learn More
Chemglass Life Sciences Plate glass, square, borosilicate, 8in X 8in X 1/8
Supplier: Chemglass Life Sciences CG190421
Plate glass, square, borosilicate, 8in X 8in X 1/8. Pyrex or Duran borosilicate plate glass. The 1/16in, 1/8in and 1/4in thick plates have edges that are cut and swiped. The 1/2in thick and over have ground edges.