Learn More
Biotang Inc 8 x10 in Autoradiography Film,cassettes Blue 800 Speed.Feature lightweight aluminum construction, sponge backing for screen-film contact, self-locking push buttons, No ID Window
Supplier:  Biotang Inc AFC810B
Encompass_Preferred
8 x10 in Autoradiography Film,cassettes Blue 800 Speed.Feature lightweight aluminum construction, sponge backing for screen-film contact, self-locking push buttons, No ID Window
Catalog No. 50-751-4992