Learn More
Andwin Scientific 8X10 BL BSC AUTORAD FLM 100/PK
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Andwin Scientific F90238X10
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The 8" x 10" GeneMate blue autoradiography film is ideal for different applications like the chemiluminescence and autoradiography of 32P and 125I. This product lasts 2 years from the date of the manufacturing. It is sold in a box of 100.