Learn More
Penn Valve And Fitting Co 8OZ LIQUID LEAK DETECTOR
Supplier: Penn Valve And Fitting Co MSSNOOP8OZ
Detector, Leak, For pressurized gas systems, Swagelok Snoop, 8 oz. liquid, Nontoxic/Noncorrosive/Nonflammable, Temp. range 27degree to 200degreeF , Flexible tube for hard-to-reach areas, Usable on vertical surfaces