Learn More
Thermo Scientific 842312051961 2.0MM CERAMIC
Supplier: Thermo Scientific 842312051961
Centre Tube Ceramic 2mm ID (EMT Torch); Centre tube intended for the analysis of samples containing HF acid.; Can only be used with the centre tube holder for the EMT torch.