Learn More
Cole-Parmer Oakton Conductivity and TDS Standard, 80 mS; 500 mL
Supplier: Cole-Parmer 0065332
• KCl, NaCl and 442 conversions printed on the bottle for ease of use (excluded on 60-mL bottle, but included in insert) • Temperature corrections are printed on the bottle, with certified values listed in tables and referenced to 25°C (excluded on 60-mL bottle, but included in insert) • Manufactured under ISO 9000 quality guidelines and traceable to NIST standards • If unopened, shelf life is 1 year. If opened recommended shelf life is 4 months • Conductivity options range from 1 µS/cm to 200,000 µS/cm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.