Learn More
Ace Glass, Inc. 800ML DIST APPR C 800Ml Dist Appr C
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 655315
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Complete distillation apparatus generally used for the determination of ammonia in water. Joints are standard taper 24/40.