Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™

Click to view available options
Quantity:
250 mg
1018828-72-2
C9H5F3N2O2
230.15
MFCD22682817,MFCD09995161
QRXSLIVFFDRFEB-UHFFFAOYSA-N
8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylic acid, 8-trifluoromethyl imidazo-1,2-a pyridine-2-carboxylic acid, 8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylicacid, 8-trifluoromethyl-imidazo 1,2-a pyridine-2-carboxylic acid
59443885
8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
OC(=O)C1=CN2C=CC=C(C2=N1)C(F)(F)F
1018828-72-2
C9H5F3N2O2
230.15
MFCD22682817,MFCD09995161
QRXSLIVFFDRFEB-UHFFFAOYSA-N
8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylic acid, 8-trifluoromethyl imidazo-1,2-a pyridine-2-carboxylic acid, 8-trifluoromethyl imidazo 1,2-a pyridine-2-carboxylicacid, 8-trifluoromethyl-imidazo 1,2-a pyridine-2-carboxylic acid
59443885
8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
OC(=O)C1=CN2C=CC=C(C2=N1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point ∽242°C (decomposition)
Quantity 250 mg
Formula Weight 230.15 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 8-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures