Learn More
Ace Glass, Inc. 7mm OD x 135mm porosity D filter stick
SDP
Supplier:  Ace Glass, Inc. 943524
Encompass_Preferred
7mm O.D. x 135mm gas dispersion tube for fine dispersion. Porous tip is approximately 7mm O.D. x 10mm, and fits a #7 Ace-Thred. Supplied with porosity D (10-20 micron) filter stick.
Catalog No. 50-976-773