Learn More
Chemglass Life Sciences 75L Process Reactor, Complete, Air Motor, 87.3" OAH with Motor, 18.4" Clearance Below Valve, Supplied Complete With Manifold and Shielding
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG1970675
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
75L Process Reactor, Complete, Air Motor, 87.3" OAH with Motor, 18.4" Clearance Below Valve, Supplied Complete With Manifold and Shielding. Complete reactor system on Chemglass's innovative Kwik Mount support frame. Lexan safety shields surround the reactor with the front panel hinged for easy access. Rugged 1" Stainless Steel Pipe (1.315" OD) Uprights and Heavy Duty PTFE Coated Aluminum Fittings Provide Maximum Strength and Chemical Durability. Reactor Jacket Graduation Scale on Both Sides. PLEASE NOTE: REACTOR STAND OVERALL HEIGHT WITH MOTOR AND CLEARANCE BELOW VALVE, ARE CALCULATED USING THE CG-2077-H STIRRER BEARING.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.